3-Chloro-4-nitropyridine-1-oxide 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-nitropyridine N-oxide

A232060-10G A232060-1G A232060-250MG A232060-5G
77113-032EA 96.81 USD
77113-032 77113-034 77113-036 77113-038
3-Chloro-4-nitropyridine-1-oxide 95%
3-Chloro-4-nitropyridine-1-oxide

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MW: 174.54 g/mol
MDL Number: MFCD00233948
CAS Number: 76439-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR