[Ru(bpm)3][Cl]2 98%

Supplier: Ambeed
Warning

A231719-100MG A231719-1G A231719-250MG
77113-018EA 60.77 USD
77113-018 77113-020 77113-022
[Ru(bpm)3][Cl]2 98%
[Ru(bpm)3][Cl]2

Precaución: for Research Use Only

Formula: C₂₄H₁₈ClN₁₂Ru
MW: 611.00 g/mol
MDL Number: MFCD28166514
CAS Number: 65034-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR