2-[(S)-1-Hydroxyethyl]pyridine 97%

Supplier: Ambeed
Warning

A231695-250MG A231695-5G A231695-1G
77113-014EA 29.86 USD
77113-014 77113-016 77685-637
2-[(S)-1-Hydroxyethyl]pyridine 97%
2-[(S)-1-Hydroxyethyl]pyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD06795465
CAS Number: 59042-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR