PF-573228 98%

Supplier: Ambeed
Warning

Synonyms: 6-((4-((3-(Methylsulphonyl)benzyl)amino)-5-(trifluoromethyl)pyrimidin-2-yl)amino)-3,4-dihydroquinolin-2(1H)-one

A230913-100MG A230913-1MG A230913-50MG A230913-5MG
77112-990EA 427.44 USD
77112-990 77112-992 77112-994 77112-996
PF-573228 98%
PF-573228

Precaución: for Research Use Only

Formula: C₂₂H₂₀F₃N₅O₃S
MW: 491.4861 g/mol
MDL Number: MFCD11519967
CAS Number: 869288-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR