Gimeracil 98%

Supplier: Ambeed
Warning

A230901-100MG A230901-1G A230901-5G A230901-25G A230901-100G
77112-986EA 5.15 USD
77112-986 77112-988 77600-844 77615-922 77685-634
Gimeracil 98%
Gimeracil

Precaución: for Research Use Only

Formula: C₅H₄ClNO₂
MDL Number: MFCD08458352
CAS Number: 103766-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR