3,4-Bis(benzyloxy)benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dibenzyloxybenzaldehyde

A230502-100G A230502-25G A230502-5G
77112-978EA 46.68 USD
77112-978 77112-980 77112-982
3,4-Bis(benzyloxy)benzaldehyde 98%
3,4-Bis(benzyloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₃
MW: 318.3658 g/mol
MDL Number: MFCD00004776
CAS Number: 5447-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR