5-Methylpyridine-2-sulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-pyridinesulphonamide

A230397-10G A230397-1G A230397-25G A230397-5G
77112-966EA 125.19 USD
77112-966 77112-968 77112-970 77112-972
5-Methylpyridine-2-sulfonamide 97%
5-Methylpyridine-2-sulfonamide

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
MDL Number: MFCD00661363
CAS Number: 65938-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR