1-(2-Chloropyrimidin-4-yl)piperidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A230054-1G A230054-250MG
77112-958EA 41.2 USD
77112-958 77112-960
1-(2-Chloropyrimidin-4-yl)piperidine-4-carboxylic acid 95%
1-(2-Chloropyrimidin-4-yl)piperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClN₃O₂
MDL Number: MFCD11040198
CAS Number: 1208087-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR