6-Chloro-2-pyridinecarboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloropyridine-2-carboxaldehyde, 6-Chloro-2-formylpyridine, 6-Chloropicolinaldehyde

A229990-10G A229990-1G A229990-25G A229990-5G A229990-250MG
77112-950EA 39.14 USD
77112-950 77112-952 77112-954 77112-956 77685-627
6-Chloro-2-pyridinecarboxaldehyde 97%
6-Chloro-2-pyridinecarboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₄ClNO
MDL Number: MFCD09832941
CAS Number: 54087-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR