α-Bromo-2,3-dichlorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: alpha-Bromo-2,3-dichlorotoluene, 2,3-Dichlorobenzyl bromide, 1-(Bromomethyl)-2,3-dichlorobenzene

A229765-10G A229765-5G
77112-944EA 66.84 USD
77112-944 77112-946
α-Bromo-2,3-dichlorotoluene 98%
α-Bromo-2,3-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
MDL Number: MFCD03701317
CAS Number: 57915-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR