H-D-Chg-OH·HCI 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Chg-OH.HCl

A229429-10G A229429-25G A229429-5G
77112-938EA 13.8 USD
77112-938 77112-940 77112-942
H-D-Chg-OH·HCI 97%
H-D-Chg-OH·HCI

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂·HCl
MW: 193.7 g/mol
MDL Number: MFCD00067543
CAS Number: 61367-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR