4-Aminobenzhydrazide 98%

Supplier: Ambeed
Warning

Synonyms: 4-Aminobenzoic hydrazide, 4-Aminobenzene-1-carbohydrazide, 4-Aminobenzoic acid hydrazide, 4-Aminobenzohydrazide

A229414-10G A229414-5G A229414-25G
77112-934EA 18.04 USD
77112-934 77112-936 77685-626
4-Aminobenzhydrazide 98%
4-Aminobenzhydrazide

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MW: 151.17 g/mol
MDL Number: MFCD00007606
CAS Number: 5351-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR