2,3,4-Trifluorobenzoic acid 98%

Supplier: Ambeed
Warning

A228294-100G A228294-10G A228294-1G A228294-25G A228294-5G
77112-896EA 81.69 USD
77112-896 77112-898 77112-900 77112-902 77112-904
2,3,4-Trifluorobenzoic acid 98%
2,3,4-Trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
MDL Number: MFCD00061232
CAS Number: 61079-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR