1-Chloro-5-fluoro-2-iodobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-fluoro-1-iodobenzene, 2-Chloro-4-fluoroiodobenzene

A227140-100G A227140-10G A227140-25G A227140-500G A227140-5G
77112-870EA 80.34 USD
77112-870 77112-872 77112-874 77112-876 77112-878
1-Chloro-5-fluoro-2-iodobenzene 97%
1-Chloro-5-fluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFI
MW: 256.45
MDL Number: MFCD00051774
CAS Number: 101335-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR