1,1,3,3-Tetramethylguanidine 97%

Supplier: Ambeed
Danger

Synonyms: N,N,N',N'-Tetramethylguanidine

A226881-10G A226881-25G
77112-868EA 5.31 USD
77112-868 77550-466
1,1,3,3-Tetramethylguanidine 97%
1,1,3,3-Tetramethylguanidine

Precaución: for Research Use Only

Formula: C₅H₁₃N₃
MW: 115.18 g/mol
MDL Number: MFCD00008323
CAS Number: 80-70-6
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR