2-Chloro-5-fluoro-6-methylpyridine 97%

Supplier: Ambeed
Warning

A226655-10G A226655-25G A226655-5G A226655-1G A226655-100G
77112-862EA 25.76 USD
77112-862 77112-864 77112-866 77593-466 77685-619
2-Chloro-5-fluoro-6-methylpyridine 97%
2-Chloro-5-fluoro-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MDL Number: MFCD04972391
CAS Number: 884494-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR