1,2-Diamino-4-chloro-5-fluorobenzene 97%
Supplier: Ambeed
|
Warning
|
A226242-10G
A226242-1G
A226242-25G
A226242-5G
77112-842EA
33.99
USD
77112-842
77112-844
77112-846
77112-848
77112-850
1,2-Diamino-4-chloro-5-fluorobenzene 97%
1,2-Diamino-4-chloro-5-fluorobenzene
Precaución: for Research Use Only
Formula:
C₆H₆ClFN₂ |
MDL Number:
MFCD00042485 CAS Number: 139512-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...