6-Bromo-2,3,4,9-tetrahydro-1H-carbazol-1-one 95%

Supplier: Ambeed
Warning

A225824-1G A225824-25G A225824-5G
77112-834EA 32.97 USD
77112-834 77112-836 77112-838
6-Bromo-2,3,4,9-tetrahydro-1H-carbazol-1-one 95%
6-Bromo-2,3,4,9-tetrahydro-1H-carbazol-1-one

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MDL Number: MFCD00459403
CAS Number: 59514-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR