3-Amino-3-(4-methoxyphenyl)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-3-(4-methoxyphenyl)propionic Acid, 3-Amino-3-(4-methoxyphenyl)propanoic acid

A224483-1G A224483-250MG A224483-5G A224483-10G
77112-814EA 11.34 USD
77112-814 77112-816 77112-818 77685-609
3-Amino-3-(4-methoxyphenyl)propionic acid 97%
3-Amino-3-(4-methoxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00187209
CAS Number: 5678-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR