2-Bromoanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-methoxybenzene

A223996-1000G A223996-100G A223996-10G A223996-25G A223996-500G
77112-804EA 164.44 USD
77112-804 77112-806 77112-808 77112-810 77112-812
2-Bromoanisole 98%
2-Bromoanisole

Precaución: for Research Use Only

Formula: C₇H₇BrO
MW: 187.04 g/mol
MDL Number: MFCD00000064
CAS Number: 578-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR