(T-4)-Bis[6-methoxy-2-[(S)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl-κO]-1H-benzimidazolato-κN3]magnesium 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Esomeprazole Magnesium
A223103-100MG
A223103-1G
A223103-250MG
A223103-25MG
A223103-50MG
A223103-5G
A223103-25G
77112-784EA
9.55
USD
77112-784
77112-786
77112-788
77112-790
77112-792
77112-794
77685-600
(T-4)-Bis[6-methoxy-2-[(S)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl-κO]-1H-benzimidazolato-κN3]magnesium 97%
(T-4)-Bis[6-methoxy-2-[(S)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl-κO]-1H-benzimidazolato-κN3]magnesium
Precaución: for Research Use Only
Formula:
C₃₄H₃₆MgN₆O₆S₂ MW: 713.12 g/mol |
MDL Number:
MFCD06798050 CAS Number: 161973-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...