3-Chloro-2,5-dimethylpyrazine 97%

Supplier: Ambeed
Warning

A221180-10G A221180-1G A221180-25G A221180-100G
77112-758EA 23.68 USD
77112-758 77112-760 77112-762 77685-593
3-Chloro-2,5-dimethylpyrazine 97%
3-Chloro-2,5-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MDL Number: MFCD00006143
CAS Number: 95-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR