4-Bromo-2,5-xylenol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,5-dimethylphenol

A220882-100G A220882-10G A220882-1G A220882-250MG A220882-25G A220882-5G
77112-746EA 1641.22 USD
77112-746 77112-748 77112-750 77112-752 77112-754 77112-756
4-Bromo-2,5-xylenol 97%
4-Bromo-2,5-xylenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.0600 g/mol
MDL Number: MFCD00029746
CAS Number: 85223-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR