3,5-Dimethoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3,5-Dimethoxyphenyl)methanol

A220455-10G A220455-25G
77112-742EA 14.42 USD
77112-742 77112-744
3,5-Dimethoxybenzyl alcohol 98%
3,5-Dimethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
MDL Number: MFCD00004641
CAS Number: 705-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR