4,4-Dimethyl-2-cyclohexen-1-one 97%

Supplier: Ambeed
Warning

A220396-100G A220396-10G A220396-1G A220396-25G A220396-5G
77112-726EA 178.19 USD
77112-726 77112-728 77112-730 77112-732 77112-734
4,4-Dimethyl-2-cyclohexen-1-one 97%
4,4-Dimethyl-2-cyclohexen-1-one

Precaución: for Research Use Only

Formula: C₈H₁₂O
MDL Number: MFCD00009695
CAS Number: 1073-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR