NVP-BHG712 98%

Supplier: Ambeed
Danger

Synonyms: NVP-BHG 712, 4-Methyl-3-((1-methyl-6-(pyridin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-yl)amino)-N-(3-(trifluoromethyl)phenyl)benzamide, 4-Methyl-3-[[1-methyl-6-(3-pyridinyl)-1H-pyrazolo[3,4-d]pyrimidin-4-yl]amino]-N-[3-(trifluoromethyl)phenyl]benzamide

A219560-10MG A219560-1MG A219560-25MG A219560-50MG A219560-5MG
77112-680EA 85.49 USD
77112-680 77112-682 77112-684 77112-686 77112-688
NVP-BHG712 98%
NVP-BHG712

Precaución: for Research Use Only

Formula: C₂₆H₂₀F₃N₇O
MDL Number: MFCD20272929
CAS Number: 940310-85-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR