3-Cyclopropylpropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Cyclopropylpropanoic acid

A217436-1G A217436-250MG
77112-642EA 182.48 USD
77112-642 77112-644
3-Cyclopropylpropionic acid 98%
3-Cyclopropylpropionic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD02179408
CAS Number: 5618-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR