2,4-Dichloro-6-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3,5-dichloro-α,α,α-trifluorotoluene, 4,6-Dichloro-α,α,α-trifluoro-o-toluidine

A217415-100G A217415-250MG A217415-5G
77112-640EA 154.5 USD
77112-640 77685-587 77685-588
2,4-Dichloro-6-(trifluoromethyl)aniline 97%
2,4-Dichloro-6-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄Cl₂F₃N
MW: 230.02
MDL Number: MFCD00174313
CAS Number: 62593-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR