tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Bromoacetyl)-1-Boc-piperazine, tert-Butyl 4-(2-bromoacetyl)piperazine-1-carboxylate
A217067-1G
A217067-250MG
A217067-5G
77112-622EA
210.06
USD
77112-622
77112-624
77685-585
tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate 95%
tert-Butyl-4-(bromoacetyl)piperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₉BrN₂O₃ MW: 307.19 g/mol |
MDL Number:
MFCD16990579 CAS Number: 112257-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...