z-trp(Boc)-OH·DCHA 95%

Supplier: Ambeed
Warning

Synonyms: Z-Trp(Boc)-OH·DCHA, Cbz-Trp(Boc)-OH·DCHA, Z-Trp(Boc)-OH.DCHA

A216799-10G A216799-1G A216799-25G A216799-5G A216799-100G
77112-594EA 37.08 USD
77112-594 77112-596 77112-598 77112-600 77568-098
z-trp(Boc)-OH·DCHA 95%
z-trp(Boc)-OH·DCHA

Precaución: For research use only.

Formula: C₃₆H₄₉N₃O₆
MW: 619.803 g/mol
MDL Number: MFCD01318754
CAS Number: 218938-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR