3-Chloro-2-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-2-methoxyphenyl)boronic acid

A216396-100MG A216396-1G A216396-250MG A216396-5G A216396-25G A216396-100G
77112-586EA 9.27 USD
77112-586 77112-588 77112-590 77112-592 77593-410 77685-584
3-Chloro-2-methoxyphenylboronic acid 97%
3-Chloro-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₃
MW: 186.4006 g/mol
MDL Number: MFCD08277278
CAS Number: 179898-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR