1-((Benzyloxy)carbonyl)-3-((tert-butoxycarbonyl)amino)piperidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A215757-100MG A215757-1G A215757-250MG A215757-5G
77112-554EA 54.11 USD
77112-554 77112-556 77112-558 77550-396
1-((Benzyloxy)carbonyl)-3-((tert-butoxycarbonyl)amino)piperidine-3-carboxylic acid 95%
1-((Benzyloxy)carbonyl)-3-((tert-butoxycarbonyl)amino)piperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₂₆N₂O₆
MW: 378.41 g/mol
MDL Number: MFCD04114485
CAS Number: 885270-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR