2,4,6-Trimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A215102-100G A215102-10G A215102-1G A215102-25G A215102-5G
77112-540EA 70.03 USD
77112-540 77112-542 77112-544 77112-546 77112-548
2,4,6-Trimethoxybenzaldehyde 98%
2,4,6-Trimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00003313
CAS Number: 830-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR