4-Bromo-4'-methoxybenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)(4-methoxyphenyl)methanone

A214881-250MG A214881-5G
77112-536EA 43.5 USD
77112-536 77112-538
4-Bromo-4'-methoxybenzophenone 98%
4-Bromo-4'-methoxybenzophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO₂
MW: 291.14 g/mol
MDL Number: MFCD00228667
CAS Number: 54118-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR