3-Methyl-2-thiophenecarbonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 3-Methylthiophene-2-carbonitrile

A214291-1G A214291-5G
77112-530EA 14.86 USD
77112-530 77112-532
3-Methyl-2-thiophenecarbonitrile 98%
3-Methyl-2-thiophenecarbonitrile

Precaución: for Research Use Only

Formula: C₆H₅NS
MW: 123.18 g/mol
MDL Number: MFCD00052086
CAS Number: 55406-13-8
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR