NSC-319726 98%

Supplier: Ambeed
Warning

Synonyms: NSC319726, N'-(1-(Pyridin-2-yl)ethylidene)azetidine-1-carbothiohydrazide

A213189-10MG A213189-1MG A213189-50MG A213189-5MG
77112-500EA 32.97 USD
77112-500 77112-502 77112-504 77112-506
NSC-319726 98%
NSC-319726

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₄S
MDL Number: MFCD01665446
CAS Number: 71555-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR