5-Formyl-2-methoxybenzoic acid 96%
Supplier: Ambeed
|
Warning
|
A213143-100MG
A213143-1G
A213143-5G
77112-496EA
5.15
USD
77112-496
77112-498
77685-566
5-Formyl-2-methoxybenzoic acid 96%
5-Formyl-2-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₈O₄ |
MDL Number:
MFCD01442588 CAS Number: 84923-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...