Quercitrin 98%

Supplier: Ambeed
Warning

A212853-1MG A212853-5MG A212853-100MG A212853-1G A212853-250MG
77112-492EA 6.18 USD
77112-492 77112-494 77685-562 77685-563 77685-564
Quercitrin 98%
Quercitrin

Precaución: for Research Use Only

Formula: C₂₁H₂₀O₁₁
MDL Number: MFCD00016932
CAS Number: 522-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR