2-Fluoro-6-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-picoline

A211927-100G A211927-10G A211927-5G A211927-25G A211927-500G
77112-470EA 88.06 USD
77112-470 77112-472 77112-474 77568-044 77685-556
2-Fluoro-6-methylpyridine 97%
2-Fluoro-6-methylpyridine

Precaución: For research use only.

Formula: C₆H₆FN
MW: 111.12 g/mol
MDL Number: MFCD00041226
CAS Number: 407-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR