4-Amino-3-methylpyridine 97%

Supplier: Ambeed
Warning

A211816-100G A211816-25G A211816-1G
77112-466EA 225.57 USD
77112-466 77112-468 77685-555
4-Amino-3-methylpyridine 97%
4-Amino-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂
MDL Number: MFCD01704431
CAS Number: 1990-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR