2-Bromo-α,α,α,6-tetrafluorotoluene 97%

Supplier: Ambeed
Warning

A211739-100G A211739-10G A211739-1G A211739-25G A211739-5G
77112-456EA 181.28 USD
77112-456 77112-458 77112-460 77112-462 77112-464
2-Bromo-α,α,α,6-tetrafluorotoluene 97%
2-Bromo-α,α,α,6-tetrafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
MDL Number: MFCD01631569
CAS Number: 261951-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR