Zoledronic acid monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy-2-(1-imidazolyl)ethane-1,1-diphosphonic acid monohydrate, Reclast, Zoledronate monohydrate, ZOL 446 monohydrate, NDC-zoledronate monohydrate, CGP42446A monohydrate, (1-Hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl)diphosphonic acid monohydrate, (1-Hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl)diphosphonic acid hydrate, Zometa

A211103-1MG A211103-50MG A211103-5G
77112-452EA 5.31 USD
77112-452 77112-454 77615-786
Zoledronic acid monohydrate 98%
Zoledronic acid monohydrate

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O₈P₂
MW: 290.10 g/mol
MDL Number: MFCD08448695
CAS Number: 165800-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR