5-Fluoro-2-methoxynicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxy-5-fluoro-2-methoxypyridine, 5-Fluoro-2-methoxypyridine-3-carboxylic acid

A210864-100MG A210864-1G A210864-250MG A210864-5G
77112-444EA 6.18 USD
77112-444 77112-446 77112-448 77112-450
5-Fluoro-2-methoxynicotinic acid 98%
5-Fluoro-2-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MDL Number: MFCD04972399
CAS Number: 884494-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR