4-(4-Hydroxy-3-iodophenyl)benzonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 4'-Hydroxy-3'-iodo-[1,1'-biphenyl]-4-carbonitrile

A210589-1G A210589-250MG A210589-5G
77112-428EA 36.05 USD
77112-428 77112-430 77112-432
4-(4-Hydroxy-3-iodophenyl)benzonitrile 95%
4-(4-Hydroxy-3-iodophenyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₈INO
MDL Number: MFCD06796628
CAS Number: 460746-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR