4-(3-Azetidinyl)morpholine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Azetidin-3-yl)morpholine dihydrochloride

A210378-5G A210378-1G
77112-426EA 225.98 USD
77112-426 77685-548
4-(3-Azetidinyl)morpholine dihydrochloride 97%
4-(3-Azetidinyl)morpholine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆Cl₂N₂O
MW: 215.12 g/mol
MDL Number: MFCD11617823
CAS Number: 178312-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR