6-Amino-1,4-benzodioxane 96%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxin-6-amine, 1,4-Benzodioxan-6-amine

A209738-1G A209738-25G
77112-422EA 14.42 USD
77112-422 77615-764
6-Amino-1,4-benzodioxane 96%
6-Amino-1,4-benzodioxane

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD00006824
CAS Number: 22013-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR