Ketanserin 95%

Supplier: Ambeed
Warning

Synonyms: 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2,4(1H,3H)-quinazolinedione, 3-(2-(4-(4-Fluorobenzoyl)piperidin-1-yl)ethyl)quinazoline-2,4(1H,3H)-dione

A209484-100MG A209484-1MG A209484-50MG A209484-250MG
77112-398EA 51.5 USD
77112-398 77112-400 77112-402 77612-054
Ketanserin 95%
Ketanserin

Precaución: for Research Use Only

Formula: C₂₂H₂₂FN₃O₃
MDL Number: MFCD00083392
CAS Number: 74050-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR