2-(4-((5,6-Diphenylpyrazin-2-yl)(isopropyl)amino)butoxy)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: MRE-269

A209226-10MG A209226-1MG A209226-25MG A209226-50MG A209226-5MG A209226-1G A209226-100MG A209226-250MG
77112-388EA 12.36 USD
77112-388 77112-390 77112-392 77112-394 77112-396 77615-756 77685-541 77685-542
2-(4-((5,6-Diphenylpyrazin-2-yl)(isopropyl)amino)butoxy)acetic acid 98%
2-(4-((5,6-Diphenylpyrazin-2-yl)(isopropyl)amino)butoxy)acetic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₉N₃O₃
MDL Number: MFCD11976884
CAS Number: 475085-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR