Flutamide 98%

Supplier: Ambeed
Danger

A208282-100G A208282-10G A208282-25G A208282-5G
77112-374EA 97.84 USD
77112-374 77112-376 77112-378 77112-380
Flutamide 98%
Flutamide

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃N₂O₃
MDL Number: MFCD00072009
CAS Number: 13311-84-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR