3-Amino-5-bromo-2-fluoropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-fluoropyridin-3-amine

A208229-10G A208229-1G A208229-5G
77112-368EA 206.88 USD
77112-368 77112-370 77112-372
3-Amino-5-bromo-2-fluoropyridine 95%
3-Amino-5-bromo-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MW: 191.0011 g/mol
MDL Number: MFCD06659524
CAS Number: 884495-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR